Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV strain A2 Rabbit anti-Virus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA123413

Catalog No. PA123413


Add to cart

Description

Description

PA1-23413 detects Respiratory Syncytial Virus (A2 Strain) in Virus samples. PA1-23413 has been successfully used in ELISA and Western Blot procedures. The PA1-23413 immunogen corresponds to Human Respiratory Syncytial Virus (RSV) Glycoprotein (G) protein of the A2 strain. Store at 4°C for short term. For extended storage aliquot and store at -20°C or below. Avoid freeze/thaw cycles.

Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Specifications

Specifications

RSV strain A2
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
human respiratory syncytial virus; RSV; RSV A2
Human Respiratory Syncytial Virus (RSV) Glycoprotein (G) protein of the A2 strain
Affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit