Learn More
RSV M2 Rabbit anti-Virus, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA123415
Catalog No. PIPA123415
Add to cart
Description
Description
PA1-23415 detects Respiratory Syncytial Virus M2 Protein in Virus samples. PA1-23415 has been successfully used in ELISA and Western Blot procedures. The PA1-23415 immunogen corresponds to Human Respiratory Syncytial Virus (RSV) M2 protein. The epitope has been mapped to residues L(81) E S Y I G S I N N I T K Q S A(95) of the human RSV M2 protein, which is a CTL epitope. 93% identity to bovine RSV M2.
Specifications
Specifications
RSV M2
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
human respiratory syncytial virus; RSV; RSV M2; RSV Matrix protein 2
Human Respiratory Syncytial Virus (RSV) M2 protein
Affinity chromatography
RUO
Antibody
Polyclonal