Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV M2 Rabbit anti-Virus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14183

Catalog No. PA14183


Add to cart

Description

Description

PA1-4183 detects Respiratory Syncytial Virus M2 Protein from infected samples. PA1-4183 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~20 kDa protein representing RSV M2 in infected HEp-2 cells. The PA1-4183 immunogen is a synthetic peptide corresponding to residues L(81) E S Y I G S I N NI T K Q S A(95) of the human RSV M2 protein, which is a CTL epitope. This sequence is 93% conserved in bovine.

Specifications

Specifications

RSV M2
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
Virus
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
human respiratory syncytial virus; RSV; RSV M2; RSV Matrix protein 2
Synthetic peptide corresponding to residues L(81) E S Y I G S I N NI T K Q S A(95) of the human RSV M2 protein, which is a CTL epitope.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit