Learn More
RGR Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00149W
Catalog No. PIOSR00149W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Receptor for all-trans- and 11-cis-retinal. Binds preferentially to the former and may catalyze the isomerization of the chromophore by a retinochrome-like mechanism.
Specifications
Specifications
RGR
Polyclonal
whole serum with no preservative
Q9Z2B3
RPE-retinal G protein-coupled receptor, Rgr
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Rgr
Lyophilized
RGR
A synthetic peptidefrom mouse retinal G protein coupled receptor
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
57811