Learn More
Rb Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA138555
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is normal colon or colon carcinoma.
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia. Phosphorylation of Rb at S608 depends on CDK4.
Specifications
| Rb | |
| Polyclonal | |
| RB1 | |
| Retinoblastoma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5925 | |
| 4° C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| P06400 | |
| RB1 | |
| Synthetic peptide derived from C-terminus of human retinoblastoma | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.