Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RARB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138562

Catalog No. 01-673-705


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is testis or heart tissue.

Retinoids are metabolites of vitamin A (retinal) and are believed to represent important signaling molecules during vertebrate development and tissue differentiation. Two families of retinoid receptors have been identified. Retinoic acid receptors (RARs) include RARalpha, RARBeta and RARgamma, each of which has a high affinity for all-trans retinoic acids and belongs to the same class of nuclear transcription factors as thyroid hormone receptors, vitamin D3 receptor and ecdysone receptor. The ligand binding domains of the RARs are highly conserved and RAR isoforms are expressed in distinct patterns throught development and in the mature organism. Members of the retinoid X receptor (RXR) family, RXRalpha, RXRBeta and RXRgamma, are activated by 9-cis-retinoic acid, a stereoand photoisomer of all-trans-RA that is expressed in vivo in both liver and kidney and may represent a widely used hormone.
Specifications

Specifications

RARB
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P10826
RAR-b
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5915
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RARB
Liquid
RARB
Synthetic peptide derived from N-terminus of human RAR beta
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit