Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RXRB Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA19097
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
RXRB Polyclonal antibody specifically detects RXRB in Human samples. It is validated for Western Blot.Specifications
RXRB | |
Polyclonal | |
Unconjugated | |
Rxrb | |
AL023085; DADB-100D22.1; DAUDI6; H-2RIIBP; MGC1831; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; NR2B2; nuclear receptor coregulator 1; nuclear receptor co-regulator 1; nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; RCoR-1; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; retinoid X receptor; retinoid X receptor beta; retinoid X receptor, beta; Rub; RXR Beta; Rxrb; RXR-beta | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P28702 | |
Rxrb | |
Internal sequence corresponding to amino acids CRDGMGDSGRDSRSP. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction