Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rev-erb A alpha/NR1D1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP219574

Catalog No. NBP219574


Add to cart

Description

Description

Rev-erb A alpha/NR1D1 Polyclonal specifically detects Rev-erb A alpha/NR1D1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rev-erb A alpha/NR1D1
0.79mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
P20393
EAR-1, EAR1nuclear receptor subfamily 1 group D member 1, HREV, nuclear receptor subfamily 1, group D, member 1, Rev-erbA-alpha, Rev-ErbAalpha, THRA1, THRALrev-erbA-alpha, V-erbA-related protein 1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
9572
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300
Affinity Purified
NR1D1
Recombinant protein encompassing a sequence within the center region of human NR1D1. The exact sequence is proprietary.
67 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only