Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rex-1/ZFP42, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131948

Catalog No. NBP131948

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Rex-1/ZFP42 Polyclonal antibody specifically detects Rex-1/ZFP42 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Rex-1/ZFP42
0.83mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
ZFP42
Recombinant protein encompassing a sequence within the center region of human Rex1. The exact sequence is proprietary.
35 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
132625
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only