Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC2, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA126567

Catalog No. PIPA126567

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also called activator 1, is a protein complex consisting of five distinct subunits of 145, 40, 38, 37, and 36.5 kD. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP. Deletion of this gene has been associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described.

PA1-26567 detects RFC2 from human samples. PA1-26567 will not react with mouse.PA1-26567 has been successfully used in Western blot procedures.The PA1-26567 immunogen is a synthetic peptide corresponding to a portion of human Replication Factor C 2 40kDa subunit encoded within exon 1.
Specifications

Specifications

RFC2
Polyclonal
Unconjugated
RFC2
Purified
RFC2
Synthetic peptide corresponding to a portion of human Replication Factor C 2 40 kDa subunit encoded within exon 1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1mg/mL
tris citrate, tris phosphate with 0.1% sodium azide; pH 7-8
P35250
A1, MGC3665, RFC40
Goat
IgG
50μL
Store at 4°C
Primary
5982
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit