Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC3, Polyclonal, Invitrogen™

Catalog No. PIPA133150
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA133150 50 μg
1 options

Catalog No. PIPA133150

Supplier: Thermo Scientific PA133150

Rabbit Polyclonal Antibody

The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins proliferating cell nuclear antigen (PCNA) and activator 1. The elongation of primed DNA templates by DNA polymerase delta and DNA poly

PA1-33150 detects RFC3 from human samples.PA1-33150 has been successfully used in Western blot applications.The PA1-33150 immunogen is: Synthetic peptide corresponding to a region with human RFC3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC3
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene RFC3
Gene Accession No. P40938
Gene Alias MGC5276, RFC38
Gene Symbols RFC3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region with human RFC3.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5983
Target Species Human
Content And Storage Store at 4°C or Store at -20°C if preferred
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.