Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523596

Catalog No. PIPA523596

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RFT1
Polyclonal
PBS with 0.09% sodium azide
Q96AA3
CDG1N, RFT1, requiring fifty three 1 homolog, protein RFT1 homolog, putative endoplasmic reticulum multispan transmembrane protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91869
Immunohistochemistry, Western Blot
Unconjugated
RFT1
Liquid
RFT1
KLH conjugated synthetic peptide between 513-541 amino acids from the C-terminal region of human RFT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit