Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFXANK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14119

Catalog No. PA14119


Add to cart

Description

Description

PA1-4119 detects RFX-B in human samples. PA1-4119 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~40 kDa protein representing RFX-B in Raji lymphoblastoid cell line. The PA1-4119 immunogen is a synthetic peptide corresponding to residues A(18) S E L G D P E D P G E E A C(31) of human RFX-B.

RFX-B is the gene responsible for the bare lymphocyte syndrome (BLS) which is characterized by a lack of MHC class II transcription. Subsequently, there is a lack of B and T cell responses to foreign antigens. RFX-B is a MHC class II transcription factor that is defective in BLS.
Specifications

Specifications

RFXANK
Polyclonal
Unconjugated
RFXANK
Liquid
RFXANK
Synthetic peptide corresponding to residues A(18) S E L G D P E D P G E E A C(31) of human RFX-B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O14593
DNA-binding protein RFXANK, Regulatory factor X subunit B, Ankyrin repeat family A protein 1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8625
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit