Learn More
anti-RFX1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP220142
Catalog No. NBP220142
Add to cart
Description
Description
RFX1 Polyclonal specifically detects RFX1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
RFX1
1mg/mL
Western Blot 1:500-1:3000
P22670
EFC, EF-C, Enhancer factor C, MHC class II regulatory factor RFX, MHC class II regulatory factor RFX1, Regulatory factor X 1, regulatory factor X, 1 (influences HLA class II expression), RFX, trans-acting regulatory factor 1, Transcription factor RFX1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
5989
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
RFX1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human RFX1. The exact sequence is proprietary.
105 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human