Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RFXANK, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB600205

 View more versions of this product

Catalog No. NB600205


Add to cart

Description

Description

RFXANK Polyclonal specifically detects RFXANK in Human, Primate samples. It is validated for Western Blot.
Specifications

Specifications

RFXANK
1 mg/mL
Western Blot 1:100-1:2000
Affinity Purified
RFXANK
Peptide made to a portion of the RFX-B protein.
42 kDa
0.1 ml
Adaptive Immunity, Immunology
Primary
8625
Human, Primate
Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
ANKRA1Regulatory factor X subunit B, ankyrin repeat-containing regulatory factor X-associated protein, BLS, DNA-binding protein RFXANK, F14150_1, MGC138628, regulatory factor X-associated ankyrin-containing proteinAnkyrin repeat family A protein 1, RFXB, RFX-BRFX-Bdelta4
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reacts with residues 18-31 [ASELGDPEDPGEEAC] of the RFX-B protein.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only