Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regulator of G-protein signaling 1 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RGS1 protein.

Manufacturer:  Abnova Corporation H00005996B02

Catalog No. 89-006-143


Add to cart

Description

Description

This gene encodes a member of the regulator of G-protein signalling family. This protein is located on the cytosolic side of the plasma membrane and contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signalling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. [provided by RefSeq

Sequence: MPGMFFSANPKELKGTTHSLLDDKMQKRRPKTFGMDMKAYLRSMIPHLESGMKSSKSKDVLSAAEVMQWSQSLEKLLANQTGQNVFGSFLKSEFSEENIEFWLACEDYKKTESDLLPCKAEEIYKAFVHSDAAKQINIDFRTRESTAKKIKAPTPTCFDEAQKVIYTLMEKDSYPRFLKSDIYLNLLNDLQANSLK
Specifications

Specifications

regulator of G-protein signaling 1
Unconjugated
No additive
NM_002922
1R20|BL34|IER1|IR20
Mouse
50 uL
Signal Transduction
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human RGS1 protein.
RGS1
Serum
RGS1
RGS1 (NP_002913, 1 a.a. ~ 196 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5996
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit