Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141146

Catalog No. 01-675-723


Add to cart

Description

Description

Suggested positive control: Jurkat cell lysate.

Regulators of G-protein signaling (RGSs) are a protein family containing a conserved 120 amino acid domain (RGS domain), which bind directly to activated G-alpha subunits (Gai and Gaq) and act as GTPase-activating proteins (GAPs). RGS1 shows a high degree of selectivity to regulate alpha2A-adrenoreceptor-activated G(o1)alpha rather than G(i1)alpha, G(i2)alpha or G(i3)alpha and different capacities to inactivate this G protein.
Specifications

Specifications

RGS1
Polyclonal
Unconjugated
RGS1
Liquid
RGS1
Synthetic peptide corresponding to residues 1-50 of human RGS1.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q08116
1R20
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5996
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit