Learn More
anti-RGS11, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP131212
Catalog No. NBP131212
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
RGS11
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
RGS11
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human RGS11. The exact sequence is proprietary.
53 kDa
100 ul
Signal Transduction
Primary
8786
Human
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
regulator of G-protein signaling 11, regulator of G-protein signalling 11, RS11
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Bovine (81%). Not yet tested in any other species.