Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS17, Polyclonal, Invitrogen™

Catalog No. PIPA125695
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA125695 50 μg
1 options

Catalog No. PIPA125695

Supplier: Thermo Scientific PA125695

Chicken Polyclonal Antibody

This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.

PA1-25695 detects RGS17 / RGSZ2 from human, mouse, and rat samples. PA1-25695 has been successfully used in Western blot procedures. It has a predicted molecular weight of 25 kDa.The PA1-25695 immunogen is a synthetic peptide corresponding to residues 41-100 of RGS17.Store at 4ºC short term. For extended storage, store at -20ºC, avoiding freeze/thaw cycles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS17/RGSZ2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene RGS17
Gene Accession No. Q9UGC6, Q9QZB0
Gene Alias RGS-17, RGSZ2, hRGS17
Gene Symbols RGS17
Host Species Chicken
Immunogen Synthetic peptide corresponding to residues 41-100 of RGS17.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26575
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.