Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS19 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523210

Catalog No. PIPA523210


Add to cart

Description

Description

RGS19 is a member of the RGS protein family, which acts as a GTPase-activating protein (GAPs) for heterotrimeric G proteins, increasing the intrinsic rate of GTP hydrolysis and accelerating signal termination. It is a small protein that contains two potential phosphorylation sites for protein kinase C and seven for casein kinase 2. The gene for RGS19 is localized on human chromosome 20q13.33. RGS19 specifically controls signaling of the Wnt-catenin pathway. The protein is highly expressed in human lung, liver, heart and placenta.
Specifications

Specifications

RGS19
Polyclonal
Unconjugated
RGS19
Liquid
RGS19
Synthetic peptide (4 PHEAEKQITGPEEADRPPS 22) of RGS19 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P49795, Q9CX84
GAIP, RGSGAIP, G alpha interacting protein, G protein signalling regulator 19, G-alpha-interacting protein, guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein, regulator of G-protein signalling 19
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10287, 56470
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit