Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS19 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528407

Catalog No. PIPA528407


Add to cart

Description

Description

PA5-28407 targets RGS19 in WB applications and shows reactivity with Human samples. The PA5-28407 immunogen is synthetic peptide corresponding to a region within amino acids 155 and 217 of Human RGS19. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

G proteins mediate a number of cellular processes. The protein encoded by this gene belongs to the RGS (regulators of G-protein signaling) family and specifically interacts with G protein, GAI3. This protein is a guanosine triphosphatase-activating protein that functions to down-regulate Galpha i/Galpha q-linked signaling. Alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.
Specifications

Specifications

RGS19
Polyclonal
Unconjugated
RGS19
Liquid
RGS19
Synthetic peptide corresponding to a region within amino acids 155 and 217 of Human RGS19
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
P49795
GAIP, RGSGAIP, G alpha interacting protein, G protein signalling regulator 19, G-alpha-interacting protein, guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein, regulator of G-protein signalling 19
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10287
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit