Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056736
Description
RGS2 Polyclonal specifically detects RGS2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RGS2 | |
Polyclonal | |
Unconjugated | |
PBS, 0.2% gelatin with 0.05% Sodium Azide | |
Cell growth-inhibiting gene 31 protein, G0 to G1 switch regulatory 8, 24kD, G0/G1 switch regulatory protein 8, G0S8cell growth-inhibiting protein 31, regulator of G-protein signaling 2, regulator of G-protein signaling 2, 24kDa, regulator of G-protein signalling 2, 24kD, regulator of G-protein signalling 2, 24kDa | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
Cross reacts with Human, Mouse, Rat. The amino acid sequence used is 86% homologous in mouse and rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml | |
P41220 | |
RGS2 | |
A synthetic peptide within amino acid residues 1-50 of human RGS2 was used as immunogen. | |
0.1 mg | |
Cell Cycle and Replication, Signal Transduction | |
5997 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction