Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129877

Catalog No. PA129877

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-29877 detects RGS2 from rat, human, mouse samples.PA1-29877 has been successfully used in Western blot applications.The PA1-29877 immunogen is: Full length human RGS2 (Amino acids 1-211)Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Specifications

Specifications

RGS2
Polyclonal
Unconjugated
RGS2
Purified
RGS2
Full length human RGS2 (Amino acids 1-211)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
1mg/mL
PBS with no preservative
P41220, O08849, Q9JHX0
G0S8
Chicken
IgY
50μg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
5997
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit