Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS22 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525541

Catalog No. PIPA525541

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RGS22 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form.
Specifications

Specifications

RGS22
Polyclonal
PBS with 0.09% sodium azide
Q8NE09, G3UYX5
PRTD-NY2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26166, 626596
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
RGS22
Liquid
RGS22
KLH conjugated synthetic peptide between 388-417 amino acids from the central region of human RGS22
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit