Learn More
RGS5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523231
Catalog No. PIPA523231
Add to cart
Description
Description
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators. The mRNA of the human gene is abundantly expressed in heart, lung, skeletal muscle, and small intestine, and at low levels in brain, placenta, liver, colon, and leukocytes.
Specifications
Specifications
RGS5
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
O15539
RP11-430G6.2, MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
RGS5
Liquid
RGS5
Amino acids 56-67 CDEALQWRDSLDK of human RGS5 were used as immunogen.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8490