Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056659SS
Description
RGS6 Polyclonal specifically detects RGS6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RGS6 | |
Polyclonal | |
Unconjugated | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
DKFZp313G1241, FLJ43552, G protein signaling 6 regulator, GAP, GTPase activating protein, H_DJ0283M22.1, H_DJ1108A12.1, MGC142132, regulator of G protein signaling 6, regulator of G protein signalling 6, regulator of G-protein signaling 6, regulator of G-protein signalling 6, S914, WUGSC:H_DJ0283M22.1, WUGSC:H_DJ1108A12.1 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
An approx. 54 kDa band is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 1:200-1:1000 | |
NP_004287.3 | |
RGS6 | |
This antibody was developed against synthetic peptides corresponding to amino acids 182-198 and 410-426 of human RGS6. It will cross react with mouse (approx. 472 amino acids) and rat RGS11 (approx. 721 amino acids). | |
0.05 mL | |
Signal Transduction | |
9628 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction