Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regulator of G-protein signaling 7, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RGS7 protein.

Manufacturer:  Abnova Corporation H00006000B01

Catalog No. 89-006-146


Add to cart

Description

Description

Sequence: MAQGNNYGQTSNGVADESPNMLVYRKMEDVIARMQDEKNGIPIRTVKSFLSKIPSVFSGSDIVQWLIKNLTIEDPVEALHLGTLMAAHGYFFPISDHVLTLKDDGTFYRFQTPYFWPSNCWEPENTDYAVYLCKRTMQNKARLELADYEAESLARLQRAFARKWEFIFMQAEAQAKVDKKRDKIERKILDSQERAFWDVHRPVPGCVNTTEVDIKKSSRMRNPHKTRKSVYGLQNDIRSHSPTHTPTPETKPPTEDELQQQIKYWQIQLDRHRLKMSKVADSLLSYTEQYLEYDPFLLPPDPSNPWLSDDTTFWELEASKEPSQQRVKRWGFGMDEALKDPVGREQFLKFLESEFSSENLRFWLAVEDLKKRPIKEVPSRVQEIWQEFLAPGAPSAINLDSKSYDKTTHNVKEPGRYTFEDAQEHIYKLMKSDSYPRFIRSSAYQELLQAKKKSGNSMDRRTSFEKFAQNVGKSLTSKRLTSLAQSY
Specifications

Specifications

regulator of G-protein signaling 7
Unconjugated
No additive
NM_002924
RGS7
RGS7 (NP_002915, 1 a.a. ~ 487 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
6000
Flow Cytometry, Western Blot
Mouse polyclonal antibody raised against a full-length human RGS7 protein.
RGS7
Serum
Mouse
50 uL
Membrane Receptors
Y
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit