Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHAMM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527273

Catalog No. PIPA527273


Add to cart

Description

Description

PA5-27273 targets RHAMM in WB applications and shows reactivity with Human samples. The PA5-27273 immunogen is recombit protein fragment corresponding to a region within amino acids 370 and 643 of Human RHAMM. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications

Specifications

RHAMM
Polyclonal
Unconjugated
HMMR
Liquid
HMMR
Recombit protein fragment corresponding to a region within amino acids 370 and 643 of Human RHAMM
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.39 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O75330
HMMR, CD168, IHABP, RHAMM, hyaluro mediated motility receptor, intracellular hyaluronic acid-binding protein, receptor for hyaluro-mediated motility
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3161
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit