Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHBG Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526978

Catalog No. PIPA526978


Add to cart

Description

Description

RHBG encodes one of two non-erythroid members of the Rhesus (Rh) protein family. Non-erythroid Rh protein family members are mainly expressed in the kidney and belong to the methylammonium-ammonium permease/ammonia transporters superfamily. All Rh family proteins are predicted to be transmembrane proteins with 12 membrane spanning domains and intracytoplasmic N- and C-termini.
Specifications

Specifications

RHBG
Polyclonal
PBS with 0.09% sodium azide
Q9H310, Q8BUX5
SLC42A2, Rh type B glycoprotein, Rhesus blood group, B glycoprotein, ammonium transporter Rh type B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57127, 58176
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RHBG
Liquid
RHBG
KLH conjugated synthetic peptide between 423-450 amino acids from the C-terminal region of human RHBG
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit