Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP19 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526812

Catalog No. PIPA526812

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Members of the ARHGAP family, such as ARHGAP19, encode negative regulators of Rho GTPases (see RHOA; MIM 165390), which are involved in cell migration, proliferation, and differentiation, actin remodeling, and G1 cell cycle progression.
Specifications

Specifications

ARHGAP19
Polyclonal
PBS with 0.09% sodium azide
Q14CB8
putative RhoGAP protein, rho GTPase-activating protein 19, rho-type GTPase-activating protein 19
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84986
Flow Cytometry, Western Blot
Unconjugated
ARHGAP19
Liquid
ARHGAP19
KLH conjugated synthetic peptide between 195-224 amino acids from the central region of human RHG19
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit