Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rho (-A, -B, -C) Rabbit anti-Rat, Human, Clone: 3L74, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04822
Description
This Anti-Rho (-A Antibody, -B, -C), clone 3L74 is validated for use in western blotting for the detection of Rho (-A, -B, -C).Specifications
Rho (-A, -B, -C) | |
Monoclonal | |
Unconjugated | |
NM_001664 | |
KLH-conjugated, synthetic peptide (SYPDTDVILMCFSIDSPDSLENKK) corresponding to amino acids 73-94 of human Rho (-A, -B, -C), with two terminal lysines added to increase peptide solubility | |
Primary | |
Human, Rat | |
Supernatant |
Western Blot | |
3L74 | |
Cultured supernantant in 0.05% sodium azide | |
Rabbit | |
100 μL | |
ARH12, ARHA, H12, RHOH12, RhoA, Rho12, RHO12, RHOA | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only