Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho Mouse, Unlabeled, Clone: 55, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610990

 View more versions of this product

Catalog No. BDB610990


Add to cart

Description

Description

Members of the Ras superfamily of GTPases are distantly related to the heterotrimeric G proteins and shuttle between inactive and active GTP-bound forms. Interconversion between these forms is controlled by guanine nucleotide exchange factors (GEFs) and GTPase-activating proteins (GAPs). The subfamilies of the Ras-like proteins share 30-50% homology, each member participates in a particular set of cellular functions. The Rho (Ras homolog) proteins (A, B, & C) are highly homologous and contain the consensus amino acid sequences necessary for GDP/GTP-binding and GTPase activity. Rho proteins contain a C-terminal sequence, CAAL, whose post-translational modification regulates Rho activation and function. Rho proteins modulate the in vitro activity of PI-3-kinase, PI- 4,5-kinase, and phospholipase D. Rho is a molecular switch that regulates cell morphology and motility in response to extracellular signals. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. Based on recent testing in native ELISA with recombinant proteins to CDC42 and Rac1, we have concluded that this antibody recognizes CDC42 and Rac1. BD Pharmingen has not performed such cross-reactivity testing on other family members.

Immunofluorescence, Western Blotting

Specifications

Specifications

Rho
55
Unconjugated
Affinity Purified
Human Rho aa. 1-155
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.