Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rho (-A Mouse anti-Human, Mouse, Rat, Clone: 55, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05778
Description
Detect Rho (-A, -B, -C) using this Anti-Rho (-A Antibody, -B, -C), clone 55 validated for use in IC and western blotting.Specifications
Rho | |
Monoclonal | |
Unconjugated | |
NP_001655 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunocytochemistry, Western Blot | |
55 | |
Purified mouse monoclonal IgG1 in buffer containing BSA containing 0.09% sodium azide and 50% glycerol. | |
RHOA, ARHA, ARH12, RHO12, RHOH12, Rho12, RhoA, H12 | |
Recombinant human RhoA containing amino acids 1-155. Clone 55. | |
200 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction