Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rhodopsin, Clone: 1D4, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB5417100UL

Catalog No. NC0206809


Add to cart

Description

Description

Host Species: Mouse
Clone: 1D4
Isotype: IgG1
Cross Reactivity: Mouse, Rat, Cow, Human, Zebrafish
Immunogen: Recombinant full length protein (Cow).

Applications:

Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

Specifications

Specifications

Rhodopsin
1mg/mL
Purified
Recombinant full length protein (Cow).
100μL
Primary
Bovine, Human, Murine, Rat, Zebrafish
1D4
Unconjugated
Murine
IgG1
Neuroscience, Signal Transduction
Monoclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit