Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoD Mouse anti-Human, Clone: 55/2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA17668

Catalog No. 01-670-402


Add to cart

Description

Description

MA1-7668 detects Rh(o)D human protein from human samples. MA1-7668 has been successfully used in agglutination applications. The MA1-7668 immunogen is Rh(o)D human protein. For short term storage, place at 4°C. For long term storage, place at -80°C.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during maligt cellular transformation. Blood group related antigens represent a group of carbohydrate determits carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Specifications

Specifications

RhoD
Monoclonal
Unconjugated
RHOD
Liquid
RHOD
Rh(o)D human protein.
Protein A
RUO
Antibody
Monoclonal
Human
Agglutination
55/2
PBS with 0.01% sodium azide
O00212
RhoD
Mouse
IgM
100 μg
4°C or -80°C if preferred
Primary
29984
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit