Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RhoGDI, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152064

Catalog No. NBP152064


Add to cart

Description

Description

RhoGDI Polyclonal antibody specifically detects RhoGDI in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

RhoGDI
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
ARHGDIA
Peptide with sequence C-QLAQIAAENE corresponding to internal region according to NP_004300.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Cross reacts with Cross reacts with Mouse and Rat. Expected from sequence similarity: Human, Mouse, Dog and Cow.
ELISA, Western Blot
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
NP_004300.1
GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1
Goat
IgG
0.1 mg
Signal Transduction
Primary
396
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only