Learn More
Rhox10 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523135
Catalog No. PIPA523135
Add to cart
Description
Description
Reproductive homeobox on X chromosome 10, also known as Reproductive homeobox 10, is a recently identified nuclear protein product of a 12 X-linked novel homeobox gene family that plays an important role during postnatal testis development. The members of this family are characterized by a conserved 60-amino-acid DNA-binding homeodomain. These transcription factors not only play an important role in various developmental processes such as DNA-dependent transcriptional regulation, body-plan specification, pattern formation, and cell fate determination, but also in understanding the development and physiology of the human reproductive system.
Specifications
Specifications
Rhox10
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Liquid
RHOX10
Synthetic peptide (90 NSKKYTNAQMCELEK 104) of Rhox-10 protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
434769
Western Blot
Unconjugated
Rhox10
reproductive homeobox on X chromosome 10, reproductive homeobox on chromosome X, 10
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse