Learn More
anti-Rhox8, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP223671SS
 View more versions of this product
Catalog No. NBP223671SS
Add to cart
Description
Description
Rhox8 Polyclonal antibody specifically detects Rhox8 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
Specifications
Rhox8
Unconjugated
0.1 ml crude antibody whole antisera with 50% Glycerol with 0.025% Sodium Azide
reproductive homeobox 8, Rhox8 reproductive homeobox 8
Within the range of amino acids 30-60 of mouse Rhox-8 protein were used as the immunogen for the antibody.
Unpurified
RUO
Primary
434768
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Peptide ELISA 1:100-1:2000
Antisera
Rabbit
IgG
0.025 ml
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen for the FLII antibody is 100% homologous in human, rhesus monkey, horse, mouse, rat and chimpanzee, 94% homologous in cattle and dog, and 89% homologous in chicken.