Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNase1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528278

Catalog No. PIPA528278


Add to cart

Description

Description

PA5-28278 targets Ribonuclease A in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28278 immunogen is recombit fragment corresponding to a region within amino acids 1 and 156 of Human Ribonuclease A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the pancreatic-type of secretory ribonucleases, a subset of the ribonuclease A superfamily. The encoded endonuclease cleaves internal phosphodiester RNA bonds on the 3 ft.-side of pyrimidine bases. It prefers poly(C) as a substrate and hydrolyzes 2 ft.,3 ft.-cyclic nucleotides, with a pH optimum near 8.0. The encoded protein is monomeric and more commonly acts to degrade ds-RNA over ss-RNA. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.
Specifications

Specifications

RNase1
Polyclonal
Unconjugated
RNASE1
Liquid
RNASE1
Recombit fragment corresponding to a region within amino acids 1 and 156 of Human Ribonuclease A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P07998
RIB1, RNS1, HP-RNase, RIB-1, RNase 1, RNase A, RNase upI-1, ribonuclease 1, ribonuclease A, ribonuclease pancreatic
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6035
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit