Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RICH2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524152

Catalog No. PIPA524152


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state. Acts as a GTPase activitor in vitro for CDC42 and RAC1.
Specifications

Specifications

RICH2
Polyclonal
PBS with 0.09% sodium azide
Q17R89
NPC-A-10, RICH2, RICH-2, Rho-type GTPase-activating protein RICH2, RhoGAP interacting with CIP4 homologs protein 2, rho GTPase-activating protein 44, rho GTPase-activating protein RICH2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9912
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARHGAP44
Liquid
ARHGAP44
KLH conjugated synthetic peptide between 784-811 amino acids from the C-terminal region of human RICH2
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit