Learn More
RIG-I (NT) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061040
Catalog No. 06-104-0MI
Add to cart
Description
Description
Use Anti-RIG-I (N-Terminus) Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunoprecipitation to detect RIG-I (N-Terminus) also known as Retinoic acid-inducible gene 1 protein.
Specifications
Specifications
RIG-I (NT)
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Immunoprecipitation, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
DDX58
GST-tagged recombinant protein corresponding to the N-terminus of human RIG-I.
100μg
Primary
NP_055129.2