Learn More
RIG-I, CT Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061041
Catalog No. 06-104-1MI
Add to cart
Description
Description
Anti-RIG-I, C-terminus Antibody is an antibody against RIG-I, C-terminus for use in Western Blotting.
Specifications
Specifications
RIG-I
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
DDX58
KLH-conjugated linear peptide corresponding to the C-terminus of human RIG-I.
100μg
Primary
NP_055129