Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RILPL2 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524505

Catalog No. PIPA524505

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, rat and Xenopus based on sequence homology.

Does not regulate lysosomal morphology and distribution.
Specifications

Specifications

RILPL2
Polyclonal
PBS with 0.09% sodium azide
Q969X0
RLP2, RILP-like protein 2, p40phox-binding protein, rab-interacting lysosomal protein-like 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100774122, 196383
Immunohistochemistry, Western Blot
Unconjugated
RILPL2
Liquid
RILPL2
KLH conjugated synthetic peptide between 120-149 amino acids from the central region of human RILPL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit