Learn More
RING1 Mouse anti-Human, Clone: 2D5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519824
Catalog No. PIMA519824
Add to cart
Description
Description
Peptide Sequence: NKECPTCRKK LVSKRSLRPD PNFDALISKI YPSREEYEAH QDRVLIRLSR LHNQQALSSS IEEGLRMQAM HRAQRVRRPI PGSDQTTTMS
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.
Specifications
Specifications
RING1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q06587
RING1A, RNF1, E3 ubiquitin-protein ligase RING1, polycomb complex protein RING1, really interesting new gene 1 protein
Mouse
IgG2b, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6015
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
2D5
Unconjugated
RING1
Liquid
RING1
RING1 (NP_002922, 81 a.a. ∽ 170 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human