Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK3/RIP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 23 publications
$204.00 - $482.50
Specifications
Antigen | RIPK3/RIP3 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 3-7 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224588SS
|
Novus Biologicals
NBP224588SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP224588
|
Novus Biologicals
NBP224588 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
RIPK3/RIP3 Polyclonal specifically detects RIPK3/RIP3 in Human, Primate, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
RIPK3/RIP3 | |
Western Blot 3-7 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Rabbit | |
Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
0.1 ml PBS with 0.05% Sodium Azide | |
EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
RIPK3 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen is 100% homologous in human and chimp. This antibody is not suitable for testing for mouse RIP3. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Primate, Mouse (Negative) | |
Q9Y572 | |
11035 | |
RIPK3/RIP3 Antibody was made to a synthetic peptide (QEGPKDPEAWSRPQ) corresponding to amino acids 497-518 of human RIPK3/RIP3 was used as immunogen for this antibody (isoform CRA_a, 518 amino acids, NP_006862.2). This also corresponds to amino acids 298-311 of isoform CRA_b (GenBank: EAW66022.1). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
56.7 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title