Learn More
RIT2 Mouse anti-Human, Clone: 3F2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519822
Catalog No. PIMA519822
Add to cart
Description
Description
Peptide Sequence: MEVENEASCS PGSASGGSRE YKVVMLGAGG VGKSAMTMQF ISHQFPDYHD PTIEDAYKTQ VRIDNEPAYL DILDTAGQAE FTAMREQYMR GGEGFIICYS VTDRQSFQEA AKFKELIFQV RHTYEIPLVL VGNKIDLEQF RQVSTEEGLS LAQEYNCGFF ETSAALRFCI DDAFHGLVRE IRKKESMPSL MEKKLKRKDC LWKKLKGSLK KKRENMT
RIN belongs to the RAS (HRAS; MIM 190020) superfamily of small GTPases (Shao et al., 1999 [PubMed 10545207]).
Specifications
Specifications
RIT2
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q99578
RIBA, RIN, ROC2
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6014
ELISA, Western Blot
3F2
Unconjugated
RIT2
Liquid
RIT2
RIT2 (AAH18060, 1 a.a. ∽ 217 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human