Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125559

Catalog No. PA125559


Add to cart

Description

Description

PA1-25559 detects RIT2 from human samples. PA1-25559 has been successfully used in Western blot procedures. The PA1-25559 immunogen is recombit, His-tagged, full length human protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

RIN belongs to the RAS (HRAS; MIM 190020) superfamily of small GTPases (Shao et al., 1999 [PubMed 10545207]).
Specifications

Specifications

RIT2
Polyclonal
Unconjugated
RIT2
Liquid
RIT2
Recombit, His-tagged, full length human protein.
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.08% sodium azide; pH 7.3
Q99578
RIBA, RIN, ROC2
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6014
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit