Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RLA-DR Mouse anti-Rabbit, R-PE, Clone: RDR34, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 407009
Description
SouthernBiotech offers anti-rabbit RLA-DR and RLA-DQ antibodies for for use in a variety of applications.
Specifications
RLA-DR | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Rabbit leukemic B cell line | |
RUO | |
Rabbit | |
IgG2b |
Flow Cytometry | |
RDR34 | |
R-PE | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title