Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL51 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524414

Catalog No. PIPA524414


Add to cart

Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Pseudogenes corresponding to this gene are found on chromosomes 4p and 21q.
Specifications

Specifications

MRPL51
Polyclonal
PBS with 0.09% sodium azide
Q4U2R6, Q9CPY1
HSPC241, MRP64, bMRP64, 39S ribosomal protein L51, mitochondrial, L51mt, MRP-L51, bMRP-64, mitochondrial ribosomal protein 64, mitochondrial ribosomal protein bMRP64
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51258, 66493
Western Blot
Unconjugated
MRPL51
Liquid
MRPL51
KLH conjugated synthetic peptide between 100-128 amino acids from the C-terminal region of human RM51
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit