Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF185 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524305

Catalog No. PIPA524305

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, rat and zebrafish based on sequence homology.

Specifications

Specifications

RNF185
Polyclonal
PBS with 0.09% sodium azide
Q96GF1, Q91YT2
BSK65-MONO1, BSK65-MONO2, BSK65-PANC1, BSK65-PANC2, BSK65-TEST1, BSK65-TEST2, BSK65-TEST3, E3 ubiquitin-protein ligase RNF185
Rabbit
IgG
400 μL
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
100761198, 193670, 91445
Flow Cytometry, Western Blot
Unconjugated
RNF185
Liquid
RNF185
KLH conjugated synthetic peptide between 88-117 amino acids from the central region of human RN185
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit